Learn More
Keystone Cleanroom Products Autoclavable Breather Bag for Isolators
These self-sealing breather bags offer a chevron peel and hang hole to suspend previously steam sterilized pouches in an isolator for VHP decontamination of outer packaging.
$1385.51 - $7259.48
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Certifications/Compliance | COC |
| Closure Type | Self-seal |
| Color | White |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Length (English) | Width (English) | Capacity (Metric) | Price | Qty | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (English) | Width (English) | Capacity (Metric) | Price | Qty | |||
|
17000795
|
Keystone Cleanroom Products
BBMGTVKSS0306HH |
6 in. | 3 in. | 0.44 L |
Case of 1000 Each for $2,626.69
|
|
|||
|
17000783
|
Keystone Cleanroom Products
BBMGTVKSS02095HH |
9.5 in. | 2 in. | 0.47 L |
Case of 1000 Each for $3,088.52
|
|
|||
|
17000888
|
Keystone Cleanroom Products
BBMGTVKSS03508HH |
8 in. | 3.5 in. | 0.69 L |
Case of 500 Each for $1,472.10
|
|
|||
|
17000799
|
Keystone Cleanroom Products
BBMGTVKSS0308HH |
8 in. | 3 in. | 0.6 L |
Case of 1000 Each for $1,385.51
|
|
|||
|
17001049
|
Keystone Cleanroom Products
BBMGTVKSS0510HH |
10 in. | 5 in. | 1.42 L |
Case of 500 Each for $1,847.34
|
|
|||
|
17001050
|
Keystone Cleanroom Products
BBMGTVKSS0608HH |
8 in. | 6 in. | 1.42 L |
Case of 500 Each for $1,847.34
|
|
|||
|
17000889
|
Keystone Cleanroom Products
BBMGTVKSS0415HH |
15 in. | 4 in. | 1.49 L |
Case of 500 Each for $2,554.53
|
|
|||
|
17001048
|
Keystone Cleanroom Products
BBMGTVKSS0422HH |
22 in. | 4 in. | 1.95 L |
Case of 500 Each for $2,554.53
|
|
|||
|
17001051
|
Keystone Cleanroom Products
BBMGTVKSS0612HH |
12 in. | 6 in. | 1.98 L |
Case of 500 Each for $2,554.53
|
|
|||
|
17001056
|
Keystone Cleanroom Products
BBMGTVKSS0810HH |
10 in. | 8 in. | 1.99 L |
Case of 500 Each for $2,626.69
|
|
|||
|
17001055
|
Keystone Cleanroom Products
BBMGTVKSS0713HH |
13 in. | 7 in. | 2.26 L |
Case of 500 Each for $2,626.69
|
|
|||
|
17001057
|
Keystone Cleanroom Products
BBMGTVKSS0815HH |
15 in. | 8 in. | 2.98 L |
Case of 500 Each for $3,016.37
|
|
|||
|
17001060
|
Keystone Cleanroom Products
BBMGTVKSS1012HH |
11 in. | 10 in. | 2.98 L |
Case of 500 Each for $3,550.36
|
|
|||
|
17001058
|
Keystone Cleanroom Products
BBMGTVKSS0818HH |
18 in. | 8 in. | 3.56 L |
Case of 500 Each for $3,867.86
|
|
|||
|
17001062
|
Keystone Cleanroom Products
BBMGTVKSS1214HH |
14 in. | 12 in. | 4.2 L |
Case of 500 Each for $4,012.20
|
|
|||
|
17001052
|
Keystone Cleanroom Products
BBMGTVKSS0628HH |
28 in. | 6 in. | 4.2 L |
Case of 500 Each for $4,012.20
|
|
|||
|
17001053
|
Keystone Cleanroom Products
BBMGTVKSS0632HH |
32 in. | 6 in. | 4.8 L |
Case of 500 Each for $5,975.00
|
|
|||
|
17001061
|
Keystone Cleanroom Products
BBMGTVKSS1020HH |
20 in. | 10 in. | 4.97 L |
Case of 500 Each for $4,243.11
|
|
|||
|
17001063
|
Keystone Cleanroom Products
BBMGTVKSS1217HH |
17 in. | 12 in. | 5 L |
Case of 500 Each for $4,329.70
|
|
|||
|
17001054
|
Keystone Cleanroom Products
BBMGTVKSS0635HH |
35 in. | 6 in. | 5.2 L |
Case of 500 Each for $6,220.35
|
|
|||
|
17001059
|
Keystone Cleanroom Products
BBMGTVKSS0834HH |
34 in. | 8 in. | 6.76 L |
Case of 500 Each for $6,364.66
|
|
|||
|
17001066
|
Keystone Cleanroom Products
BBMGTVKSS1617HH |
17 in. | 16 in. | 6.76 L |
Case of 500 Each for $5,873.97
|
|
|||
|
17001065
|
Keystone Cleanroom Products
BBMGTVKSS1520HH |
20 in. | 15 in. | 7.45 L |
Case of 500 Each for $5,946.12
|
|
|||
|
17001064
|
Keystone Cleanroom Products
BBMGTVKSS1424HH |
24 in. | 14 in. | 8.25 L |
Case of 500 Each for $6,797.64
|
|
|||
|
17001070
|
Keystone Cleanroom Products
BBMGTVKSS1822HH |
22 in. | 18 in. | 9.8 L |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17001067
|
Keystone Cleanroom Products
BBMGTVKSS1626HH |
26 in. | 16 in. | 10.3 L |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17001068
|
Keystone Cleanroom Products
BBMGTVKSS1632HH |
32 in. | 16 in. | 12.7 L |
Case of 250 Each for $4,676.08
|
|
|||
|
17001069
|
Keystone Cleanroom Products
BBMGTVKSS1635HH |
35 in. | 16 in. | 13.9 L |
Case of 250 Each for $5,267.81
|
|
|||
|
17001071
|
Keystone Cleanroom Products
BBMGTVKSS2426HH |
26 in. | 24 in. | 15.5 L |
Case of 250 Each for $4,863.70
|
|
|||
|
17001072
|
Keystone Cleanroom Products
BBMGTVKSS2430HH |
30 in. | 24 in. | 17.9 L |
Case of 250 Each for $5,152.34
|
|
|||
|
17001073
|
Keystone Cleanroom Products
BBMGTVKSS2435HH |
35 in. | 24 in. | 20.9 L |
Case of 250 Each for $5,513.16
|
|
|||
Description
- Tyvek™ and polyethylene film construction for high strength and exceptional microbial barrier does not absorb or adsorb VHP
- Designed for steam sterilization, but also works with gamma irradiation, EtO and VHP
- Fiber-free peelable seals
- High clarity for product visibility
- Recommended for sterilization of pharmaceutical equipment
- Manufactured and packaged in a certified ISO 6 cleanroom
- Works with both Impulse and Continuous Band Sealers
- Improves the efficiency of Parts Preparation Department
Specifications
| Autoclavable | |
| Self-seal | |
| Single-use | |
| Tyvek | |
| 203 μm | |
| Steam, EtO, Gamma Irradiation, VHP | |
| Autoclave Bag |
| COC | |
| White | |
| Isolator | |
| Non-sterile | |
| Yes | |
| 8 mil |