missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aushon Cirasoft™ Analyst Software
Description
Easy to Use
- Optimized for accurate analysis of quantitative protein arrays
- Quickly and accurately locates and quantifies each spot within each well
- Standard protocols are easily created and saved
- Internal database provides rapid recall of standard and stored protocols
- Organizes the final data output by analyte and by sample based on user-defined assay templates, making the information much easier to process
- Auto spot location, alignment and measurement make image analysis fast and simple
- Images can be archived for a permanent record of raw experimental data
- Multiple exposures are used to form an expanded dynamic range curve to improve the measurement of low level samples while preventing the high level samples from reaching saturation levels
- Nearly instantaneous recalculation of curve fit and sample measurement
- Color heat map and gray scale options provide selectable image viewing and manipulation
Superior Design
- Simple, fast, out-of-the box installation – ready to use with minimal instruction
- Compact; uses <2 sq. ft. of bench space
- All components integrated with embedded MacMini and iPad user interface, eliminating need for separate system components
- Unlike fluidic array analysis systems, no daily, weekly or monthly maintenance is required
Specifications
Specifications
| For Use With (Application) | Analyze both small and large protein arrays |