Learn More
Aushon Chemiluminescent Soluble Human Cytokine Receptor Array Kits
For simultaneous measurement of combinations of soluble cytokine receptors
Supplier: Aushon Biosystems Inc 070061FA
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate
Specifications
| Human | |
| For simultaneous measurement of combinations of soluble Cytokine receptors | |
| 2 Plex |