Learn More
Aushon Chemiluminescent Rat Cytokine Array Kits
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Specifications
| For Use With (Application) | For simultaneous measurement of combinations of Cytokines |
| Product Type | Cytokine Array Kits |
| Quantity | 8 Plex |