Learn More
Aushon Chemiluminescent Rat Cytokine Array Kits
Catalog No. 232941fa
Encompass
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 Plex
4 Plex
6 Plex
8 Plex
Catalog No. Quantity
232941FA 4 Plex
232921FA 4 Plex
233F61FA 8 Plex
232B01FA 4 Plex
233961FA 6 Plex
233FF1FA 10 Plex
6 options
Catalog No. 232941FA
Supplier: Aushon Biosystems Inc 232941FA
Only null left
For simultaneous measurement of combinations of cytokines
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
For Use With (Application) For simultaneous measurement of combinations of Cytokines
Product Type Cytokine Array Kits
Quantity 4 Plex