Learn More
Aushon Chemiluminescent Rat Chemokine Array Kits
Catalog No. 242581FA
Encompass
Change view
Click to view available options
Quantity:
10 Plex
4 Plex
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
242581FA 4 Plex
243FF1FA 10 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 242581FA Supplier Aushon Biosystems Inc Supplier No. 242581FA
For simultaneous measurement of combinations of chemokines
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
For Use With (Application) For simultaneous measurement of combinations of chemokines
Quantity 4 Plex