Learn More
Aushon Chemiluminescent Porcine Cytokine Array Kits
Catalog No. p-4480450
Encompass
Change view
Click to view available options
Quantity:
1 Plex
3 Plex
7 Plex
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
260011FA 1 Plex
2600E1FA 3 Plex
267F01FA 7 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 260011FA Supplier Aushon Biosystems Inc Supplier No. 260011FA
Only null left
For simultaneous measurement of combinations of cytokines
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
For Use With (Application) For simultaneous measurement of combinations of cytokines
Quantity 1 Plex