Learn More
Aushon Chemiluminescent Mouse Neurotrophic Factor 1 Array Kits
Catalog No. 200011FA
Encompass
Change view
Click to view available options
Quantity:
1 Plex
Catalog No. Quantity
200011FA 1 Plex
1 options
Catalog No. 200011FA Supplier Aushon Biosystems Inc Supplier No. 200011FA
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For measurement of neurotrophic factors
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
For Use With (Application) For measurement of neurotrophic factors
Quantity 1 Plex