Learn More
Aushon Chemiluminescent Mouse MMP Array Kits
Catalog No. 200021FA
Encompass
Change view
Click to view available options
Quantity:
1 Plex
2 Plex
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
200021FA 1 Plex
200601FA 2 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 200021FA Supplier Aushon Biosystems Inc Supplier No. 200021FA
For simultaneous measurement of combinations of cytokines
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
For Use With (Application) For simultaneous measurement of combinations of cytokines
Quantity 1 Plex