Learn More
Aushon Chemiluminescent Mouse Cytokine Array Kits
For simultaneous measurement of combinations of cytokines
Supplier: Aushon Biosystems Inc 17C841FA
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
| For simultaneous measurement of combinations of cytokines | |
| Cytokine Array Kits | |
| 4 Plex |