Learn More
Aushon Chemiluminescent Mouse Cytokine Array Kits
Catalog No. 17FFF1FA
Encompass
Change view
Click to view available options
Detects:
Cytokine 1
Cytokine 2
Cytokine 3
Cytokine 4
Cytokine 5
Cytokine 6
Cytokine 7
Quantity:
1 Plex
10 Plex
12 Plex
4 Plex
7 Plex
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detects Quantity
17FFF1FA Cytokine 1 12 Plex
17C841FA Cytokine 5 4 Plex
178941FA Cytokine 4 4 Plex
17F851FA Cytokine 3 7 Plex
178851FA Cytokine 6 4 Plex
17FF51FA Cytokine 2 10 Plex
190021FA Cytokine 7 1 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17FFF1FA Supplier Aushon Biosystems Inc Supplier No. 17FFF1FA
For simultaneous measurement of combinations of cytokines
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Detects Cytokine 1
For Use With (Application) For simultaneous measurement of combinations of cytokines
Product Type Cytokine Array Kits
Quantity 12 Plex