Learn More
Aushon Chemiluminescent Mouse Chemokine Array Kits
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Specifications
| For Use With (Application) | For simultaneous measurement of combinations of chemokines |
| Product Type | Chemokine Array Kits |
| Quantity | 5 Plex |