Learn More
Aushon Chemiluminescent Mouse Chemokine Array Kits
Catalog No. 190011FA
Encompass
Change view
Click to view available options
Detects:
Chemokine 1
Chemokine 2
Chemokine 3
Chemokine 4
Quantity:
1 Plex
10 Plex
5 Plex
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detects Quantity
190011FA Chemokine 2 1 Plex
195981FA Chemokine 3 5 Plex
19A641FA Chemokine 4 5 Plex
19FFC1FA Chemokine 1 10 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 190011FA Supplier Aushon Biosystems Inc Supplier No. 190011FA
For simultaneous measurement of combinations of chemokines
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Detects Chemokine 2
For Use With (Application) For simultaneous measurement of combinations of chemokines
Product Type Chemokine Array Kits
Quantity 1 Plex