Learn More
Aushon Chemiluminescent iMac™ with Microsoft™ Office and PROArray™ Software
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Specifications
| For Use With (Equipment) | Aushon Cirascan Imaging and Analysis System |