missing translation for 'onlineSavingsMsg'
Learn More

Aushon Chemiluminescent Human MMP Array Kits

Catalog No. 117001FA
Encompass
Change view
Click to view available options
Detects:
MMP 1
MMP 2
MMP 3
MMP 4
Quantity:
1 Plex
3 Plex
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detects Quantity
117001FA MMP 1 3 Plex
110041FA MMP 3 1 Plex
110021FA MMP 4 1 Plex
110E01FA MMP 2 3 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 117001FA Supplier Aushon Biosystems Inc Supplier No. 117001FA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For simultaneous measurement of matrix metalloproteinase targets

Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles

Specifications

Reactivity Human
Detects MMP 1
For Use With (Application) For simultaneous measurement of matrix metalloproteinase targets
Product Type MMP Array Kits
Quantity 3 Plex