missing translation for 'onlineSavingsMsg'
Learn More

Aushon Chemiluminescent Human MMP Array Kits

Catalog No. 110e01fa
Encompass
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Plex
3 Plex
Catalog No. Quantity
110E01FA 3 Plex
110041FA 1 Plex
110021FA 1 Plex
117001FA 3 Plex
4 options

Catalog No. 110E01FA

Supplier: Aushon Biosystems Inc 110E01FA

Only null left

For simultaneous measurement of matrix metalloproteinase targets

Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles

Specifications

Reactivity Human
For Use With (Application) For simultaneous measurement of matrix metalloproteinase targets
Product Type MMP Array Kits
Quantity 3 Plex