missing translation for 'onlineSavingsMsg'
Learn More

Aushon Chemiluminescent Human Cytokine Array Kits

Catalog No. 02FF81FA
Encompass
Change view
Click to view available options
Detects:
Cytokine 1
Cytokine 2
Cytokine 3
Cytokine 4
Cytokine 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detects
02FF81FA Cytokine 1
020071FA Cytokine 2
021981FA Cytokine 5
027681FA Cytokine 3
028981FA Cytokine 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02FF81FA Supplier Aushon Supplier No. 02FF81FA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For simultaneous measurement of combinations of cytokines

Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating cytokine profiles

Specifications

Detects Cytokine 1
For Use With (Application) For simultaneous measurement of combinations of cytokines
Product Type Cytokine Array Kits
Quantity 9 Plex
Reactivity Human