Learn More
Aushon Chemiluminescent Human Cytokine Array Kits DFS Item
For simultaneous measurement of combinations of cytokines
Supplier:  Aushon Biosystems Inc 028981FA
 View more versions of this product
Catalog No. 028981FA
Only null left
Explore available promotions
Description
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating cytokine profiles
Specifications
Specifications
Human
For simultaneous measurement of combinations of cytokines
Cytokine Array Kits
4 Plex
Product Suggestions
Product Suggestions