Learn More
Aushon Chemiluminescent Human Chemokine Array Kits
For simultaneous measurement of combinations of chemokines
Supplier: Aushon Biosystems Inc 030021FA
Description
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
| Human | |
| For simultaneous measurement of combinations of chemokines | |
| Chemokine Array Kits | |
| 1 Plex |