missing translation for 'onlineSavingsMsg'
Learn More

Aushon Chemiluminescent Human Cell Adhesion Molecule Array Kits

Catalog No. 080041FA
Encompass
Change view
Click to view available options
Quantity:
1 Plex
2 Plex
3 Plex
4 Plex
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
080041FA 1 Plex
080011FA 2 Plex
080E01FA 3 Plex
08F001FA 4 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 080041FA Supplier Aushon Biosystems Inc Supplier No. 080041FA

For simultaneous measurement of combinations of cell adhesion targets

Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles

Specifications

Reactivity Human
For Use With (Application) For simultaneous measurement of combinations of cell adhesion targets
Product Type Human Cell Adhesion Molecule Array Kits
Quantity 1 Plex