missing translation for 'onlineSavingsMsg'
Learn More

Aushon Chemiluminescent Human Angiogenesis Array Kits

Catalog No. 090E01FA
Encompass
Change view
Click to view available options
Quantity:
1 Plex
3 Plex
4 Plex
Detects:
FGF basic, HGF, PDGF-BB, VEGF
HB-EGF
PDGF-AA
PlGF, TPO, VEGF-R2
VEGF-R1
VEGF-R3
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Detects
090E01FA 3 Plex PlGF, TPO, VEGF-R2
09F001FA 4 Plex FGF basic, HGF, PDGF-BB, VEGF
090101FA 1 Plex VEGF-R3
090041FA 1 Plex PDGF-AA
090021FA 1 Plex HB-EGF
090081FA 1 Plex VEGF-R1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 090E01FA Supplier Aushon Biosystems Inc Supplier No. 090E01FA

For simultaneous measurement of combinations of angiogenesis targets

Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles

Specifications

Reactivity Human
Detects PlGF, TPO, VEGF-R2
For Use With (Application) For simultaneous measurement of combinations of angiogenesis targets
Product Type Angiogenesis Array Kits
Quantity 3 Plex