missing translation for 'onlineSavingsMsg'
Learn More

Aurintricarboxylic acid

Catalog No. p-7022919
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1590506 5 g
AAA1590514 25 g
AAA1590522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1590506 Supplier Thermo Scientific Chemicals Supplier No. A1590506

CAS: 4431-00-9 | C22H14O9 | 422.345 g/mol

Aurintricarboxylic acid is employed as a powerful inhibitor of cellular processes, including apoptosis, that are dependent on the formation of protein-nucleic acid complexes. It binds to a FGF and reduces its angiogenic activity. Aurintricarboxylic Acid has been shown to inhibit apoptotic cell death in various cell types induced by a variety of factors. It Inhibits protein-nucleic acid interactions. It stimulates the tyrosine phosphorylation of MAP kinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aurintricarboxylic acid is employed as a powerful inhibitor of cellular processes, including apoptosis, that are dependent on the formation of protein-nucleic acid complexes. It binds to a FGF and reduces its angiogenic activity. Aurintricarboxylic Acid has been shown to inhibit apoptotic cell death in various cell types induced by a variety of factors. It Inhibits protein-nucleic acid interactions. It stimulates the tyrosine phosphorylation of MAP kinases.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 4431-00-9
Molecular Formula C22H16O9
Molecular Weight (g/mol) 424.36
MDL Number MFCD00011663
InChI Key YETAHXYTPZCNBG-UHFFFAOYNA-N
Synonym aurintricarboxylic acid, aluminon free acid, aluminon, 5,5'-3-carboxy-4-oxocyclohexa-2,5-dienylidenemethylene di salicylic acid, benzoic acid, 5-3-carboxy-4-hydroxyphenyl 3-carboxy-4-oxo-2,5-cyclohexadien-1-ylidene methyl-2-hydroxy, 1,4-cyclohexadiene-1-carboxylic acid, 3-bis 3-carboxy-4-hydroxyphenyl methylene-6-oxo, 3,3'-3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene methylene bis 6-hydroxybenzoic acid, 5,5'-3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene methylene bis 2-hydroxybenzoic acid
PubChem CID 2259
ChEBI CHEBI:87397
IUPAC Name 5-[(3-carboxy-4-hydroxyphenyl)(3-carboxy-4-oxocyclohexa-2,5-dien-1-yl)methyl]-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC(C=CC1=O)C(C1=CC=C(O)C(=C1)C(O)=O)C1=CC=C(O)C(=C1)C(O)=O

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 2228904
Solubility Information Insoluble in water.
Formula Weight 422.35
Chemical Name or Material Aurintricarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-628-1
RTECSNumber GU4790000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only