Learn More
Aurintricarboxylic acid
Description
Aurintricarboxylic acid is employed as a powerful inhibitor of cellular processes, including apoptosis, that are dependent on the formation of protein-nucleic acid complexes. It binds to a FGF and reduces its angiogenic activity. Aurintricarboxylic Acid has been shown to inhibit apoptotic cell death in various cell types induced by a variety of factors. It Inhibits protein-nucleic acid interactions. It stimulates the tyrosine phosphorylation of MAP kinases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aurintricarboxylic acid is employed as a powerful inhibitor of cellular processes, including apoptosis, that are dependent on the formation of protein-nucleic acid complexes. It binds to a FGF and reduces its angiogenic activity. Aurintricarboxylic Acid has been shown to inhibit apoptotic cell death in various cell types induced by a variety of factors. It Inhibits protein-nucleic acid interactions. It stimulates the tyrosine phosphorylation of MAP kinases.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 25 g |
| Beilstein | 2228904 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 422.35 |
| Chemical Name or Material | Aurintricarboxylic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 224-628-1 |
| RTECSNumber | GU4790000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only