missing translation for 'onlineSavingsMsg'
Learn More

Auramine 80.0+%, TCI America™

Catalog No. A033425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A033425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A033425G Supplier TCI America Supplier No. A033425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2465-27-2
Molecular Formula C17H22ClN3
Molecular Weight (g/mol) 303.834
MDL Number MFCD00012484
InChI Key KSCQDDRPFHTIRL-UHFFFAOYSA-N
Synonym auramine o, basic yellow 2, auramin, auramine yellow, auramine hydrochloride, aizen auramine, auramine extra, auramine pure, auramine n, auramine chloride
PubChem CID 17170
ChEBI CHEBI:51876
IUPAC Name 4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride
SMILES CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl

Specifications

Quantity 25 g
UN Number 2811
Formula Weight 303.83
Percent Purity ≥80.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Auramine
EINECSNumber (5)-1917
RTECSNumber BY3675000
TSCA Yes