Learn More
Mystaire™ Aura™ Elite Series Ductless Fume Hood Accessory, HEPA Filters
For Aura Elite Series 30, 42 and 54 in. Ductless Fume Hoods. Provides operator safety from potentially harmful airborne particulate.
$1,421.81 - $1,685.23
Specifications
Filter Type HEPA
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
15338454
View Documents Promotion Details
Mystaire™
MY2820
Aura Elite 42 Ductless Fume Hood
Each for $1,685.23
Only null left
15338436
View Documents Promotion Details
Mystaire™
MY5920
Aura Elite 42 Ductless Fume Hood (Safety)
Each for $1,421.81
Only null left
Description
Description
  • Provides 99.97% efficient down to 0.3μ particles
Specifications
Specifications
HEPA