missing translation for 'onlineSavingsMsg'
Learn More

Attune™ Shutdown Solution (1X)

$78.00 - $253.00

Specifications

Product Line Attune
Content And Storage Store at room temperature.
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Shutdown Solution
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
501034962
Encompass
View Documents Promotion Details
N/A 250 mL
Each for $125.25
Only null left
 
A24975
View Documents Promotion Details
N/A 250 mL
Each for $78.00
Only null left
 
J106628
View Documents Promotion Details
N/A 1 L
Each for $253.00
Only null left
 
Description

Description

Attune™ Shutdown Solution is a cleaning buffer that is used in the automated shutdown procedure for the Attune™ Acoustic Focusing Cytometer.

Specifications

Specifications

Attune
Store at room temperature.
Attune Acoustic Focusing Cytometer
Shutdown Solution
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.