missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ Shutdown Solution (1X)

Catalog No. J106628
Change view
Click to view available options
Quantity:
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
J106628 1 L
501034962 250 mL
A24975 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. J106628 Supplier Invitrogen™ Supplier No. J106628

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Attune™ Shutdown Solution (1X)

  • A cleaning buffer that is used in the automated shutdown procedure for the Attune™ Acoustic Focusing Cytometer
  • Azide-free shutdown solution has been formulated to minimize bubble formulation that can occur in the system lines overnight
  • Less bubbles help keep the fluid lines clean, and minimize microbial growth in the fluid lines
  • Use at the end of each day to ensure that all sample fluids and dyes have been removed from the system
  • Ensure that fluidic lines and pumps are filled with Attune™ Shutdown Solution to prevent salt deposits from forming in the lines
  • Shutdown takes approximately 20 minutes, but most of the steps are automatic and under the control of the Attune™ Cytometric Software
  • Shutdown procedures should be run at least once a day, even if the instrument is intended for continuous use
  • Proper cleaning of the instrument ensures its consistent and accurate operation
  • The ready-to-use 1X shutdown solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system
  • The Attune™ Shutdown Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality
  • Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer

Specifications

Product Line Attune
Content And Storage Store at room temperature.
Quantity 1 L
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Shutdown Solution
I've run out of Attune Shutdown Solution for the Attune NxT Flow Cytometer. What can I use for the time being in place of the shutdown solution until I can get more shutdowns solution?

In an emergency, we recommend using the highest purity of water available in place of the shutdown solution and then switching to the Attune Shutdown Solution as soon as possible.

For Research Use Only. Not for use in diagnostic procedures.