Learn More
Invitrogen™ Attune™ NxT Violet 6 Conversion Kit
The Attune NxT Violet 6 Conversion kits are designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet.
Supplier: Invitrogen™ A36569
Description
Attune NxT Violet 6 Conversion kits are for use in the reconfiguration of four-channel Attune NxT instruments to enable detection of six channels of violet. A conversion kit is required for each original and any upgraded lasers of the instrument. The conversion is carried out in your laboratory by a Field Service Engineer.
Chemical Identifiers
| Attune | |
| Room Temperature | |
| Conversion Kit |
| Optical block and filters | |
| Attune NxT Four-Channel Violet Instrument |
Specifications
| Attune | |
| 1 kit | |
| Room Temperature | |
| Conversion Kit |
| Optical block and filters | |
| Violet | |
| Attune NxT Four-Channel Violet Instrument |
For Research Use Only. Not for use in diagnostic procedures.