missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Violet 6 Conversion Kit

Catalog No. p-200113522
Change view
Click to view available options
Color:
Blue
Red
Violet
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
A35428 Blue
A36569 Violet
A36571 Red
A36572 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A35428 Supplier Invitrogen™ Supplier No. A35428

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Attune NxT Violet 6 Conversion kits are designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet.

Attune NxT Violet 6 Conversion kits are for use in the reconfiguration of four-channel Attune NxT instruments to enable detection of six channels of violet. A conversion kit is required for each original and any upgraded lasers of the instrument. The conversion is carried out in your laboratory by a Field Service Engineer.

Specifications

Product Line Attune
Content And Storage Optical block and filters
Quantity 1 Kit
Color Blue
Shipping Condition Room Temperature
For Use With (Equipment) Attune NxT Four-Channel Violet Instrument
Product Type Conversion Kit

For Research Use Only. Not for use in diagnostic procedures.