accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Protein Analysis Reagents
Crosslinking, Labeling and Protein Modification
Protein Labeling Reagents
Atto 633 NHS Ester BioReagent, Suitable for Fluorescence, 90% (Coupling to Amines), MilliporeSigma™ Supelco™
Print
Atto 633 NHS Ester BioReagent, Suitable for Fluorescence, 90% (Coupling to Amines), MilliporeSigma™ Supelco™
Catalog No.
111003941
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
111003941
1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
111003941
Supplier
Merck Emd Millipore
Supplier No.
014641MGF
Sign In
or
Register
to check your price and availability.
Quantity
Only null left
Add to cart
Add to cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Specifications
Quantity
1 mg
Content And Storage
-20°C
Product Suggestions
Documents