missing translation for 'onlineSavingsMsg'
Learn More

Atropine 99.0+%, TCI America™

Catalog No. A07545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07545G 5 g
A075425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A07545G Supplier TCI America Supplier No. A07545G

Chemical Identifiers

CAS 51-55-8
Molecular Formula C17H23NO3
Molecular Weight (g/mol) 289.375
MDL Number MFCD00022622
InChI Key RKUNBYITZUJHSG-PJPHBNEVSA-N
Synonym r-atropine, +-atropine, +-hyoscyamine, hyoscyamine, tropan-3alpha-yl 2r-3-hydroxy-2-phenylpropanoate, 3-endo-8-methyl-8-azabicyclo 3.2.1 oct-3-yl 2r-3-hydroxy-2-phenylpropanoate, atropine sulfate, unii-v3203si72m, 1s,5r-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 2r-3-hydroxy-2-phenylpropanoate, prestwick0_000233
PubChem CID 174174
IUPAC Name [(1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate
SMILES CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3

Specifications

Melting Point 116°C
Color White
pH 10.0 (0.0015 mol H2O soln.)
Quantity 5 g
UN Number 1544
Formula Weight 289.38
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Atropine
RTECSNumber CK0700000
TSCA Yes