accessibility menu, dialog, popup

UserName

CH3NOOOH

CAS RN 51-55-8

IUPAC Name: 8-methyl-8-azabicyclo[3.2.1]octan-3-yl 3-hydroxy-2-phenylpropanoate
Synonyms: ()-hyoscyamine atropine, its salts and derivatives
Molecular Weight (g/mol): 289.38
Molecular Formula: C17H23NO3
InChi Key: RKUNBYITZUJHSG-UHFFFAOYNA-N
SMILES: CN1C2CCC1CC(C2)OC(=O)C(CO)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Product Type 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results