missing translation for 'onlineSavingsMsg'
Learn More

Atropine Sulfate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019325
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019325 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019325 Supplier Merck Emd Millipore Supplier No. PHR13791G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Atropine Sulfate is an alkaloid extracted from belladonna herb. It is a cholinergic drug, administered as an anti-arrhythmic medicine. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 5908-99-6
Physical Form Neat
Linear Formula (C17H23 NO3)2 · H2 SO4 · H2O
MDL Number MFCD00074815
Melting Point 189°C to 192°C (A)
Synonym Atropine sulfate salt monohydrate; alpha-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1 ]oct-3-yl ester; Tropine tropate
Type Certified Reference Material
Quantity 1 g
Formula Weight 694.83
Grade Certified Reference Material