missing translation for 'onlineSavingsMsg'
Learn More

Atropine Sulfate Monohydrate 99.0+%, TCI America™

Catalog No. p-7116958
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A05505G 5 g
A055025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A05505G Supplier TCI America Supplier No. A05505G
Only null left

Chemical Identifiers

CAS 5908-99-6
Molecular Formula C17H27NO8S
Molecular Weight (g/mol) 405.462
MDL Number MFCD00077268
InChI Key PVGPXGKNDGTPTD-IJTOKZDFSA-N
Synonym atropine sulfate hydrate, atropine sulfate monohydrate, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 3-hydroxy-2-phenylpropanoate; methane; sulfuric acid
PubChem CID 23624044
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate
SMILES CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.O.OS(=O)(=O)O

Specifications

Melting Point 194°C
Color White
Quantity 5 g
UN Number 1544
Formula Weight 676.82
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Atropine Sulfate Monohydrate
EINECSNumber (5)-3898
RTECSNumber CK2450000
TSCA Yes