accessibility menu, dialog, popup

UserName

CH3NOOOHH2OOHSOOHO

CAS RN 5908-99-6

IUPAC Name: 8-methyl-8-azabicyclo[3.2.1]octan-3-yl 3-hydroxy-2-phenylpropanoate sulfuric acid hydrate
Synonyms: ()-hyoscyamine sulfuric acid hydrate
Molecular Weight (g/mol): 405.46
Molecular Formula: C17H27NO8S
InChi Key: PVGPXGKNDGTPTD-UHFFFAOYNA-N
SMILES: O.OS(O)(=O)=O.CN1C2CCC1CC(C2)OC(=O)C(CO)C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)