missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atropine sulfate monohydrate, 97+%
Description
Antagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Antagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.
Solubility
Soluble in water (2500 mg/ml) at 4°C, ethanol (200 mg/ml) at 4°C, Glycerol (400 mg/ml), and methanol.
Notes
Light Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Antagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.
Solubility
Soluble in water (2500 mg/ml) at 4°C, ethanol (200 mg/ml) at 4°C, Glycerol (400 mg/ml), and methanol.
Notes
Light Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 187°C to 192°C |
| Quantity | 10 g |
| UN Number | UN1544 |
| Beilstein | 6109275 |
| Sensitivity | Light sensitive |
| Merck Index | 14,875 |
| Solubility Information | Soluble in water (2500mg/ml) at 4°C,ethanol (200mg/ml) at 4°C,Glycerol (400mg/ml),and methanol. |
| Formula Weight | 694.84 (676.82 Anhydrous) |
| Percent Purity | ≥98% |
| Chemical Name or Material | Atropine sulfate monohydrate |
Safety and Handling
| Hazard Category | H300+H330-H500 |
| Hazard Statement | GHS H Statement H300-H330 Fatal if swallowed. Fatal if inhaled. |
| Precautionary Statement | P260-P264b-P270-P271-P285-P301+P310-P304+P340-P310-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S. |
| EINECSNumber | 200-235-0 |
| RTECSNumber | CK2455000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only