missing translation for 'onlineSavingsMsg'
Learn More

Atropine sulfate monohydrate, 97+%

Catalog No. p-3777344
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226680100 10 g Glass bottle
AC226680250 25 g Glass bottle
AC226681000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC226680100 Supplier Thermo Scientific Chemicals Supplier No. 226680100

Atropine sulfate monohydrate, CAS # 5908-99-6, is a muscarinic receptor antagonist. | CAS: 5908-99-6 | H2SO4·H2O | 694.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Atropine is a competitive and reversible antagonist of muscarinic acetylcholine receptors on both central and peripheral neurons
  • Atropine was originally isolated from the belladonna plant Atropa belladonna but is also found in other Solanaceae plants
  • Atropine is a drug that can be used to slow heart rate, decrease salivary secretions, and treat poisoning caused by cholinergic or muscarinic agents

Applications

  • It is used to block activation of muscarinic receptors in laboratory mice
  • It can be used in a laboratory setting to decrease oral and airway secretions in research animals
  • Atropine has been used to study heart rate and blood pressure variability in swine

Chemical Identifiers

CAS 5908-99-6
Molecular Formula H2SO4·H2O
Molecular Weight (g/mol) 694.85
MDL Number MFCD00074815
InChI Key PVGPXGKNDGTPTD-IJTOKZDFSA-N
Synonym atropine sulfate hydrate, atropine sulfate monohydrate, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 3-hydroxy-2-phenylpropanoate; methane; sulfuric acid
PubChem CID 23624044
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate
SMILES CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.O.OS(=O)(=O)O

Specifications

Acidity Passes Test
Melting Point >187.0°C
Color White
Ignition Residue 0.2% max.
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 10 g
Beilstein 21, 27
Merck Index 15, 866
Specific Rotation − 0.60
Solubility Information Solubility in water: soluble. Other solubilities: soluble in boiling alcohol and glycerol, slightly soluble in chloroform, practically insoluble in ether
Formula Weight 694.85
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material Atropine sulfate monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Fatal if swallowed.
Causes skin irritation.
Fatal if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only