missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATRIP, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB31644
Specifications
| ATRIP | |
| Polyclonal | |
| ELISA (1:40000) Western Blot (1:500-2000) The optimal working dilution should be determined by the end user. | |
| ATR interacting protein | |
| ATRIP | |
| A synthetic peptide corresponding to amino acids 10-90 of human ATRIP (phospho S68). | |
| RUO | |
| 84126 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| In PBS (50% glycerol, 0.5% BSA and 0.02% sodium azide). | |
| DKFZp762J2115/FLJ12343/MGC20625/MGC21482/MGC26740 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of ATRIP protein only when phosphorylated at S68. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |