missing translation for 'onlineSavingsMsg'
Learn More

Atrazine, Restek

Catalog No. 06719841
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06719841 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06719841 Supplier Chromatographic Supplier No. RK 32208

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1912-24-9
Molecular Formula C8H14ClN5
Molecular Weight (g/mol) 215.69
MDL Number MFCD00041810
InChI Key MXWJVTOOROXGIU-UHFFFAOYSA-N
PubChem CID 2256
ChEBI CHEBI:15930
IUPAC Name 6-chloro-N2-ethyl-N4-(propan-2-yl)-1,3,5-triazine-2,4-diamine
SMILES CCNC1=NC(Cl)=NC(NC(C)C)=N1

Specifications

Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000ug/mL, Acetone
Chemical Name or Material Atrazine, Pesticide Mix