missing translation for 'onlineSavingsMsg'
Learn More

Atrazine, 95+%

CAS: 1912-24-9 | C8H14ClN5 | 215.69 g/mol

$170.16 - $170.16

Chemical Identifiers

CAS 1912-24-9
Molecular Formula C8H14ClN5
Molecular Weight (g/mol) 215.69
MDL Number MFCD00041810
InChI Key MXWJVTOOROXGIU-UHFFFAOYSA-N
IUPAC Name 6-chloro-N2-ethyl-N4-(propan-2-yl)-1,3,5-triazine-2,4-diamine
SMILES CCNC1=NC(Cl)=NC(NC(C)C)=N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC161680050
View Documents
Thermo Scientific Chemicals
161680050
5 g
Each for $170.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1912-24-9
215.69
MXWJVTOOROXGIU-UHFFFAOYSA-N
CCNC1=NC(Cl)=NC(NC(C)C)=N1
C8H14ClN5
MFCD00041810
6-chloro-N2-ethyl-N4-(propan-2-yl)-1,3,5-triazine-2,4-diamine

Specifications

1912-24-9
C8H14ClN5
175.0°C
Soluble in ether, chloroform and methanol, Very soluble in dimethyl sulfoxide, Insoluble in water
CCNC1=NC(Cl)=NC(NC(C)C)=N1
215.69
215.69
95+%
MFCD00041810
15, 862
MXWJVTOOROXGIU-UHFFFAOYSA-N
6-chloro-N2-ethyl-N4-(propan-2-yl)-1,3,5-triazine-2,4-diamine
5 g
Atrazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement May cause an allergic skin reaction.

May cause damage to organs through prolonged or repeated exposure.

Very toxic to aquatic life with long lasting effects.

GHS P Statement Wear protective gloves/protective clothing.

IF ON SKIN: Wash with plenty of soap and water.

If skin irritation or rash occurs: Get medical advice/attention.

Get medical attention/advice if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 217-617-8

SDS
Documents

Documents

RUO – Research Use Only