missing translation for 'onlineSavingsMsg'
Learn More

Atrazine, 95+%

Catalog No. AC161680050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC161680050 5 g
AC161680250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161680050 Supplier Thermo Scientific Chemicals Supplier No. 161680050

CAS: 1912-24-9 | C8H14ClN5 | 215.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1912-24-9
Molecular Formula C8H14ClN5
Molecular Weight (g/mol) 215.69
MDL Number MFCD00041810
InChI Key MXWJVTOOROXGIU-UHFFFAOYSA-N
IUPAC Name 6-chloro-N2-ethyl-N4-(propan-2-yl)-1,3,5-triazine-2,4-diamine
SMILES CCNC1=NC(Cl)=NC(NC(C)C)=N1

Specifications

Assay Percent Range 95+%
Melting Point 175.0°C
Merck Index 15, 862
Solubility Soluble in ether, chloroform and methanol, Very soluble in dimethyl sulfoxide, Insoluble in water
Quantity 5 g
Formula Weight 215.69
Chemical Name or Material Atrazine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement May cause an allergic skin reaction.

May cause damage to organs through prolonged or repeated exposure.

Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing.

IF ON SKIN: Wash with plenty of soap and water.

If skin irritation or rash occurs: Get medical advice/attention.

Get medical attention/advice if you feel unwell.

EINECSNumber 217-617-8

RUO – Research Use Only