missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atranol, ≥97.0% (HPLC), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 526-37-4 |
| Physical Form | Neat |
| Linear Formula | C8H8O3 |
| MDL Number | MFCD04117961 |
| Synonym | 2,6-Dihydroxy-4-methylbenzaldehyde; 4-Methyl-gamma-resorcylaldehyde; p-Orsellinaldehyde |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 152.15 |
| Percent Purity | ≥97.0% (HPLC) |
| Grade | Analytical Standard |