missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Atracurium besylate, 96%

Catalog No. p-7077887
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461470010 1 g
AC461470050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461470010 Supplier Thermo Scientific Chemicals Supplier No. 461470010
Only null left

CAS: 64228-81-5 | C53H72N2O12·2C6H5O3S | 1243.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64228-81-5
Molecular Formula C53H72N2O12·2C6H5O3S
Molecular Weight (g/mol) 1243.49
InChI Key XXZSQOVSEBAPGS-UHFFFAOYSA-L
Synonym atracurium besylate, atracurium besilate, tracrium, atracurii besilas, besilate d'atracurium, besilato de atracurio, tracrium preservative free, atracurii besilas inn-latin, bw 33a
PubChem CID 47320
ChEBI CHEBI:2915
IUPAC Name benzenesulfonate;5-[3-[1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-3,4-dihydro-1H-isoquinolin-2-ium-2-yl]propanoyloxy]pentyl 3-[1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-3,4-dihydro-1H-isoquinolin-2-ium-2-yl]propanoate
SMILES C[N+]1(CCC2=CC(=C(C=C2C1CC3=CC(=C(C=C3)OC)OC)OC)OC)CCC(=O)OCCCCCOC(=O)CC[N+]4(CCC5=CC(=C(C=C5C4CC6=CC(=C(C=C6)OC)OC)OC)OC)C.C1=CC=C(C=C1)S(=O)(=O)[O-].C1=CC=C(C=C1)S(=O)(=O)[O-]

Specifications

Melting Point 90.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (mix of isomers) (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 1243.49
Percent Purity 96%
Chemical Name or Material Atracurium besylate
EINECSNumber 264-743-4

RUO – Research Use Only