missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP6V0C Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116676
Description
Positive Samples: Mouse brain, Mouse heart, Mouse kidney, Mouse liver, Rat liver Immunogen sequence: SGTGIAAMSV MRPE.
ATP6V0C is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c', and d. ATP6V0C encodes the V0 subunit c.Chemical Identifiers
| ATP6V0C | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| ATP6V0C | |
| Atp6c; Atp6c2; Atp6l; ATP6V0C; ATPase H+ transporting V0 subunit c; ATPase, H+ transporting, lysosomal (vacuolar proton pump) 16 kDa; ATPase, H+ transporting, lysosomal (vacuolar proton pump), subunit C; ATPase, H+ transporting, lysosomal 1; ATPase, H+ transporting, lysosomal 16kD, V0 subunit C; ATPase, H+ transporting, lysosomal 16kDa, V0 subunit c; ATPase, H+ transporting, lysosomal 2; ATPase, H+ transporting, lysosomal V0 subunit C; ATPase, H+ transporting, V0 subunit C; ATPL; Atpl-rs1; H(+)-ATPase (mvp); H(+)-transporting two-sector ATPase, 16 kDa subunit; Mvp; PL16; vacuolar ATP synthase 16 kDa proteolipid subunit; vacuolar H+ ATPase proton channel subunit; vacuolar proton pump 16 kDa proteolipid subunit; VATL; V-ATPase 16 kDa proteolipid subunit; Vma3; VPPC; V-type proton ATPase 16 kDa proteolipid subunit | |
| ATP6V0C | |
| Primary | |
| Affinity chromatography |
| 1.37 mg/mL | |
| ELISA, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P27449, P63081, P63082 | |
| 11984, 170667, 527 | |
| Synthetic peptide. | |
| Antibody |
Specifications
| ATP6V0C | |
| Polyclonal | |
| Unconjugated | |
| ATP6V0C | |
| Atp6c; Atp6c2; Atp6l; ATP6V0C; ATPase H+ transporting V0 subunit c; ATPase, H+ transporting, lysosomal (vacuolar proton pump) 16 kDa; ATPase, H+ transporting, lysosomal (vacuolar proton pump), subunit C; ATPase, H+ transporting, lysosomal 1; ATPase, H+ transporting, lysosomal 16kD, V0 subunit C; ATPase, H+ transporting, lysosomal 16kDa, V0 subunit c; ATPase, H+ transporting, lysosomal 2; ATPase, H+ transporting, lysosomal V0 subunit C; ATPase, H+ transporting, V0 subunit C; ATPL; Atpl-rs1; H(+)-ATPase (mvp); H(+)-transporting two-sector ATPase, 16 kDa subunit; Mvp; PL16; vacuolar ATP synthase 16 kDa proteolipid subunit; vacuolar H+ ATPase proton channel subunit; vacuolar proton pump 16 kDa proteolipid subunit; VATL; V-ATPase 16 kDa proteolipid subunit; Vma3; VPPC; V-type proton ATPase 16 kDa proteolipid subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11984, 170667, 527 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.37 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P27449, P63081, P63082 | |
| ATP6V0C | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |