missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP6AP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537234 Supplier Invitrogen™ Supplier No. PA537234

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 52 kDa. Purity is >95% by SDS-PAGE.

Vacuolar-type H+-ATPase (V-ATPase) is a multisubunit enzyme responsible for acidification of eukaryotic intracellular organelles. V-ATPases pump protons against an electrochemical gradient, thereby synthesizing ATP. A peripheral V1 domain, which is responsible for ATP hydrolysis, and an integral V0 domain, which is responsible for proton translocation, compose the V-ATPase. Nine subunits (A-H) make up the V1 domain and five subunits (a, d, c, c' and c) make up the V0 domain. ATP6AP1 (ATPase, H+ transporting, lysosomal accessory protein 1), also known as 16A, CF2, Ac45, XAP3, ATP6S1, VATPS1 (vacuolar ATP synthase S1 accessory protein) or ATP6IP1, is a type I transmembrane, V-ATPase accessory protein that is predominantly expressed in endocrine and neuronal cells. ATP6AP1 is responsible for targeting the V-ATPase enzyme to specialized complex vacuolar systems. Via its cytoplasmic tail, ATP6AP1 interacts with subunits of the V0 domain. The disruption of this interaction in osteoclasts results in impaired bone resorption, suggesting an important role for ATP6AP1 in proper osteoclastic bone resorption.

Specifications

Antigen ATP6AP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP6AP1
Gene Accession No. O54715, Q15904, Q9R1Q9
Gene Alias 16A; Ac45; AI316502; Atp6ap1; ATP6IP1; Atp6s1; ATPase H+ transporting accessory protein 1; ATPase, H+ transporting, lysosomal (vacuolar proton pump), subunit 1; ATPase, H+ transporting, lysosomal accessory protein 1; ATPase, H+ transporting, lysosomal interacting protein 1; AW108110; C7-1; C7-1 protein; CF2; H-ATPase subunit; mFLJ00383; MGC129781; protein C7-1; Protein XAP-3; vacuolar H+ ATPase accessory subunit 1; vacuolar proton pump subunit S1; V-ATPase Ac45 subunit; V-ATPase S1 accessory protein; V-ATPase subunit Ac45; V-ATPase subunit S1; VATPS1; V-type proton ATPase subunit S1; XAP3; XAP-3
Gene Symbols ATP6AP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in human ATP6AP1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 537, 54411, 83615
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less