missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP5SL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559568 Supplier Invitrogen™ Supplier No. PA559568
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: FLTNYFYDVE ALRDYLLQRE MYKVHEKNRS YTWLEKQHGP YGAGAFFILK QGGAVKFRDK EWIRPDKYGH FSQEFWNFCE VPVEAVDAGD C Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 54%.

Required for the assembly of the mitochondrial NADH:ubiquinone oxidoreductase complex (complex I). Involved in the assembly of the distal region of complex I. [UniProt].

Specifications

Antigen ATP5SL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dmac2
Gene Accession No. Q9NW81
Gene Alias ATP synthase subunit s-like protein; ATP5S like; Atp5sl; ATP5S-like; distal membrane arm assembly complex 2; distal membrane-arm assembly complex protein 2; DMAC2
Gene Symbols Dmac2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP5SL. Recombinant protein control fragment (Product #RP-98961).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55101
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less