missing translation for 'onlineSavingsMsg'
Learn More

ATP10A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560006 Supplier Thermo Fisher Scientific Supplier No. PA560006
Only null left

ATP10A Polyclonal Antibody

ATP10A Polyclonal Antibody for IHC

Specifications

Antigen ATP10A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATP10A
Gene Accession No. O60312
Gene Alias aminophospholipid translocase VA; ATP10A; ATP10C; ATPase class V type 10A; ATPase phospholipid transporting 10A (putative); ATPase type IV, phospholipid transporting (P-type); ATPase, class V, type 10A; ATPase, Class V, type 10C; ATPVA; ATPVC; KIAA0566; P4-ATPase flippase complex alpha subunit ATP10A; phospholipid-transporting ATPase VA; probable phospholipid-transporting ATPase VA
Gene Symbols ATP10A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ATP10A. Recombinant protein control fragment (Product #RP-98196).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57194
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less