missing translation for 'onlineSavingsMsg'
Learn More

Atovaquone, Thermo Scientific™

Catalog No. p-7238590
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468762500 250 mg
AC468760010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468762500 Supplier Thermo Scientific Chemicals Supplier No. 468762500
Only null left

Chemical Identifiers

CAS 95233-18-4
Molecular Formula C22H19ClO3
Molecular Weight (g/mol) 366.84
MDL Number 00889188
InChI Key BSJMWHQBCZFXBR-UHFFFAOYSA-N
IUPAC Name 3-[4-(4-chlorophenyl)cyclohexyl]-4-hydroxy-1,2-dihydronaphthalene-1,2-dione
SMILES OC1=C(C2CCC(CC2)C2=CC=C(Cl)C=C2)C(=O)C(=O)C2=CC=CC=C12

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color Yellow to Dark Yellow
Melting Point 221°C to 225°C
Solubility Soluble in THF; Slightly soluble in acetone; Insoluble in methanol and water
Quantity 250 mg
Chemical Name or Material Atovaquone
Recommended Storage Normal conditions