missing translation for 'onlineSavingsMsg'
Learn More

Atovaquone 98.0+%, TCI America™

Catalog No. p-7117966
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2545200MG 200 mg
A25451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2545200MG Supplier TCI America Supplier No. A2545200MG

Chemical Identifiers

CAS 95233-18-4
Molecular Formula C22H19ClO3
Molecular Weight (g/mol) 366.84
MDL Number MFCD00889188
InChI Key BSJMWHQBCZFXBR-UHFFFAOYSA-N
Synonym trans-2-[4-(4-Chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthalenedione
PubChem CID 74989
IUPAC Name 3-[4-(4-chlorophenyl)cyclohexyl]-4-hydroxy-1,2-dihydronaphthalene-1,2-dione
SMILES OC1=C(C2CCC(CC2)C2=CC=C(Cl)C=C2)C(=O)C(=O)C2=CC=CC=C12

Specifications

Melting Point 223°C
Quantity 200 mg
Formula Weight 366.84
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Atovaquone
RTECSNumber QJ5616500
TSCA No
Recommended Storage Freezer