missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atovaquone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 223°C |
| Quantity | 200 mg |
| Formula Weight | 366.84 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Atovaquone |
Safety and Handling
| RTECSNumber | QJ5616500 |
| TSCA | No |
| Recommended Storage | Freezer |